Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicon carbide, beta-phase, nanopowder, 95% min

Catalog No. AA4388414
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 409-21-2 | CSi | 40.10 g/mol

Silicon carbide is used in applications requiring high endurance, such as car brakes, car clutches and ceramic plates in bulletproof vests. Used in electronic applications, measure gas temperatures in an optical technique called thin filament pyrometry. Used in jewelry, steel production, grapheme production. It is also used in production of exponential resistors, wire sawing, construction industry, a filler in plastics and composites, ceramic sintered parts, ferrites and soft metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 409-21-2
Molecular Formula CSi
Molecular Weight (g/mol) 40.10
MDL Number MFCD00049531
InChI Key HBMJWWWQQXIZIP-UHFFFAOYSA-N
Synonym silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide
PubChem CID 9863
ChEBI CHEBI:29390
SMILES [C-]#[Si+]

Specifications

Melting Point 2700°C
Color Black to Gray
Physical Form Powder
Quantity 25 g
Merck Index 14,8492
Solubility Information Insoluble in water, alcohol; Soluble in fused alkali
IUPAC Name methanidylidynesilylium
Formula Weight 40.1
Percent Purity ≥95%
Odor Odorless
Chemical Name or Material Silicon carbide, beta-phase
Show More Show Less
Hazard Category H315-H319-H335-H350
Hazard Statement GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 206-991-8
RTECSNumber VW0450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.