Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Silicon(IV) chloride, 99.998% (metals basis)
Click to view available options
Quantity:
25 mL
100 mL
Description
Silicon(IV) chloride is used as semiconductor grade.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 57.6°C |
Melting Point | -70°C |
Color | Colorless |
Physical Form | Liquid |
Quantity | 25 mL |
Linear Formula | SiCl4 |
UN Number | UN1818 |
Merck Index | 14,8500 |
Solubility Information | Reacts with water. Miscible with benzene,ether,chloroform. |
Formula Weight | 169.9 |
Show More |
Safety and Handling
Hazard Category | H301+H331-H314-H335 |
Hazard Statement | GHS H Statement H314-H335-H319 Causes severe skin burns and eye damage. May cause respiratory irritation. Causes serious eye irritation. |
Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: SILICON TETRACHLORIDE |
EINECSNumber | 233-054-0 |
RTECSNumber | VW0525000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction