Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicon(IV) chloride, 99%

Catalog No. AA69182K2
Click to view available options
Quantity:
10 mL
100 mL
1 L
3 x 1 L

CAS: 10026-04-7 | Cl4Si | 169.89 g/mol

Silicon(IV) chloride is used as an intermediate in the manufacture of polysilicon. It is also used in the semiconductors industry, photovoltaic cells and in the production of optical fibers. Its derivatives like trichlorosilane and silane are used in the Siemens process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-04-7
Molecular Formula Cl4Si
Molecular Weight (g/mol) 169.89
MDL Number MFCD00011229
InChI Key FDNAPBUWERUEDA-UHFFFAOYSA-N
Synonym silicon tetrachloride, silicon chloride, perchlorosilane, silane, tetrachloro, silicon iv chloride, tetrachlorosilicon, chlorid kremicity, silicio tetracloruro di, siliciumtetrachlorid, silicon chloride sicl4
PubChem CID 24816
IUPAC Name tetrachlorosilane
SMILES Cl[Si](Cl)(Cl)Cl

Specifications

Boiling Point 57.6°C
Melting Point °C to 70°C
Color Colorless
Physical Form Liquid
Quantity 1 L
Assay Percent Range 99% (metals basis)
Solubility Information Miscible with benzene,toluene,chloroform,petroleum ether,carbon tetrachloride,ether and hydrochloric acid.
Formula Weight 169.9
Odor Acrid
Sensitivity Moisture sensitive
Appearance Colourless liquid
Density 1.483
Chemical Name or Material Silicon(IV) chloride
Show More Show Less
Hazard Category H301+H331-H314-H335
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation DOT Class: 8, Packaging Group: II
EINECSNumber 233-054-0
RTECSNumber VW0525000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.