Learn More
Silicon(IV) chloride, 99%
Description
Silicon(IV) chloride is used as an intermediate in the manufacture of polysilicon. It is also used in the semiconductors industry, photovoltaic cells and in the production of optical fibers. Its derivatives like trichlorosilane and silane are used in the Siemens process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 57.6°C |
| Melting Point | °C to 70°C |
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 3 x 1 L |
| Assay Percent Range | 99% (metals basis) |
| Solubility Information | Miscible with benzene,toluene,chloroform,petroleum ether,carbon tetrachloride,ether and hydrochloric acid. |
| Formula Weight | 169.9 |
| Odor | Acrid |
| Sensitivity | Moisture sensitive |
| Show More |
Safety and Handling
| Hazard Category | H301+H331-H314-H335 |
| Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | DOT Class: 8, Packaging Group: II |
| EINECSNumber | 233-054-0 |
| RTECSNumber | VW0525000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.