Learn More
Silicon(IV) iodide, 99.999% (metals basis)
Description
Silicon(IV) iodide is used as a precursor to prepare silicon amides. It finds application in the manufacture and etching of silicon in microelectronics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Specifications
Specifications
| Boiling Point | 290°C |
| Melting Point | 120°C |
| Color | White |
| Physical Form | Lump |
| Quantity | 5 g |
| Linear Formula | SiI4 |
| UN Number | UN2923 |
| Solubility Information | Soluble in organic solvents. Insoluble in water. |
| Formula Weight | 535.7 |
| Percent Purity | 99.999% |
| Show More |
Safety and Handling
| Hazard Category | H301-H314-H335 |
| Hazard Statement | GHS H Statement H300-H314-H318 Fatal if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S. |
| EINECSNumber | 236-706-2 |
| TSCA | Yes |
| storageNote1 |
|
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.