Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Silodosin
GREENER_CHOICE

Catalog No. p-7161670
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective α1A adrenoceptor antagonist (Ki = 0.036 nM)

Silodosin is a selective α1A adrenoceptor antagonist (Ki = 0.036 nM). Displays >55 fold selectivity for α1A over α1B and α1D. Inhibits norepinephrine-induced [Ca2+]I increase in α1A expressing CHO cells (IC50 = 0.32 nM). Inhibits norepinephrine-induced contraction of isolated human prostate tissue.

Specifications

Chemical Name or Material 2,3-Dihydro-1-(3-hydroxypropyl)-5-[(2R)-2-[[2-[2-(2,2,2-trifluoroethoxy)phenoxy]ethyl]amino]propyl]-1H-indole-7-carboxamide
CAS 160970-54-7
Quantity 10 mg
Molecular Formula C25H32F3N3O4
Formula Weight 495.53
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.