Learn More
Thermo Scientific™ SilTite™ GC Capillary Column Connector
Catalog No. p-4522167
Click to view available options
Type:
Capillary Column Connector
Replacement Column Connector Nuts
Diameter (Metric) Inner:
0.1 to 0.25 mm
0.1 to 0.53 mm
0.45 to 0.53 mm
32 mm
Use SilTite GC Capillary Column Connectors for fused silica capillary GC columns.
Use Thermo Scientific™ SilTite Capillary Column Connectors for fused silica capillary GC columns. They are glass lined for inertness and have a low thermal mass.
Join together two GC columns
Repair broken columns
TRUSTED_SUSTAINABILITY
Specifications
Type Capillary Column Connector
Description 290 MU498
Diameter (Metric) Inner 0.1 to 0.25 mm
For Use With (Equipment) Fused Silica Capillary Column
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.