Description
- SilTite™ FingerTite technology provides easy installation and a permanent leak-tight connection
- Deactivated metal and zero-dead-volume design ensure optimal peak shapes
- Robust connection is stable through extreme temperature and pressure cycling
- When connecting columns of different inner diameters (ID), always connect the column with the largest ID first
- Ideal for use with mass spectrometers
- Includes: SilTite™ μ-Union Connectors (2), Double-Taper Ferrules (5), and installation tools
Specifications
Specifications
| For Use With | Standard Columns |
| Type | MicroUnion |
| Includes | SilTite™ μ-Union Connectors (2), Double-Taper Ferrules (5), and Installation Tools |
| For Use With (Application) | Fused Silica Analytical Columns, Guard Columns, and Retention Gaps |
| Diameter (Metric) Inner | 0.18/0.25 to 0.32 mm |