Learn More
Silver nanoparticles, 100nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 490nm absorption
Description
An alternative to gold nanoparticles due to lower absorption maximum in the 400-500nm range. Readily binds proteins and other ligands for conjugate and sensor development.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- An alternative to gold nanoparticles due to lower absorption maximum in the 400-500nm range
- Readily binds proteins and other ligands for conjugate and sensor development
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 mL |
| Odor | Odorless |
| Particle Size | 100nm |
| Sensitivity | Light sensitive |
| Form | Liquid |
| Chemical Name or Material | Silver nanoparticles, 0.02mg/mL, supplied in 2mM sodium citrate, 490nm absorption |
Safety and Handling
| TSCA | Yes |
| storageNote1 | 2° to 8°C |
| Recommended Storage | Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.