Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver sulfate-Sulfuric acid solution, Volumetric solution, for COD determination according to DIN 38409, part 41, Solstice

Catalog No. 60046385 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-385 4 x 2.5 L
60-013-57 1 L
60-048-573 6 x 1 L
60-013-58 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-046-385 Supplier Solstice Supplier No. 346294X2.5L
Add to Cart
Add to Cart

Volumetric solution, for COD determination according to DIN 38409, part 41

  • Solutions are prepared with raw materials which meet or exceed requirements of the American Chemical Society
  • Where no ACS specifications exist, chemicals of highest purity available are used
  • All aqueous solutions are prepared using high purity, deionized, water which meets requirements for ASTM type 1 Reagent Grade Water
  • Exacting production and quality control records are maintained providing traceability of raw materials
  • All solutions are guaranteed to meet label specifications
  • All solution labels meet or exceed all ISO, DOT and OSHA regulations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5 , 10294-26-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Boiling Point 338°C
CAS Min % ≥90.0000%, ≥1.0000%
CAS Max % ≤100.0000%, <5.0000%
Quantity 4 x 2.5 L
Linear Formula (CH3CH2)2O
UN Number UN3264
Packaging Glass Bottle
Density 1.85 g/cm3
Chemical Name or Material Silver sulfate-Sulfuric acid solution, Volumetric solution, for COD determination according to DIN 38409, part 41
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Corrosive liquid, acidic, inorganic, n.o.s.
Recommended Storage Room Temp
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.