Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport™ Scientific Amplitube™ PCR Reaction Strips

Catalog No. p-3747286
Click to view available options
Color:
Blue
Green
Natural
Red
Yellow
Closure Type:
Domed Cap
Flat Cap
This item is not returnable. View return policy
This item is not returnable. View return policy

Made of polypropylene. Amplitube™ PCR Reaction Strips are manufactured under the most stringent conditions to attain the highest quality standards.

  • Available with either flat (needle pierceable) or dome-topped individually attached hinged caps
  • Hinged caps have integral seal that prevents contamination of lid surface
  • Ultra-thin sidewalls and bottoms for more uniform and efficient temperature transfers
  • Positive sealing fully protects contents from evaporation during thermal cycle
  • Strip fits standard well spacing and can also be used with 8-channel hand-held pipettors
  • Easily opened and closed with one hand
  • Autoclavable
  • Choice of colorless and four non-cytotoxic and non-metallic colors
  • Packed in tamper-proof resealable bags

Specifications

Color Natural
Volume (Metric) 0.2 mL
Quantity 125 Pk.
For Use With (Equipment) Standard well spacing and can also be used with 8-channel hand-held pipettors
Sterility Non-sterile
Material Polypropylene
Closure Type Domed Cap
Autoclavable Autoclavable
Format Tube
Product Type 8-Strip PCR Reaction Tube
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.