Learn More
Description
- Made of polypropylene
- Free of RNase, DNase, Pyrogen, and DNA
- Convenient carrying handles on both sides
- Cover, grid plate, and base are keyed to prevent misalignment
- Easy to read ID numbers and letters
- Grid plate stands on 4 legs and can be placed on a lab counter or in a water bath
- These racks and tubes are also ideal for storing, freezing, and transporting reagents and specimens
Specifications
Specifications
| Color | White |
| Material | Polypropylene |
| For Use With | HTLV-III testing, bacterial and hybridoma cell uptake studies, cell harvesting |
| Includes | Rack with 12 strips of 8 tubes |
| Quantity | 10/Cs. |
| Sterility | Sterile |
| Product Type | 12-strip Tube Rack |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.