Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Simport™ Scientific Urine Collection URISAFE™ Containers
Urine Collection URISAFE™ Containers are user-friendly to both patients and laboratory personnel.
Supplier: Simport™ Scientific B3504LSP
Description
Urine Collection URISAFE™ Containers are user-friendly to both patients and laboratory personnel.
- Innovative cap features a large opening for easy collection
- A leakproof-cap liner ensures spill-free sample transport
- Vertical and horizontal graduations allow for accurate and easy reading, eliminating transfer to graduated cylinder for volume measurement
- Containers can be subjected to freezing, thawing and EtO gas sterilization without causing changes in materials or physical appearance
- Resistant to hydrochloric acid
- Convenient refrigerator storage with less wasted space
- Containers are available in 3 L and 4 L tilt size
- The 4 L bottle allows a clean and easy transfer to tubes by simply tilting the container forward without lifting
Specifications
24-Hour Urine Specimen Collection Container | |
Yes | |
9.6 x 4.52 x 7.9 in. (24.5 x 11.5 x 20.1 cm) | |
Leakproof Polypropylene screw cap, snap valve pour spout, large central handle, HCl resistant | |
4 L |
Wide-mouth Screw Cap with Integral Spout | |
HDPE | |
Spill-free sample collection and transport | |
29/Cs. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction