Learn More
Simport™ Scientific Swingsette™ Tissue Processing/Embedding Cassettes
Shop All Simport Scientific Products
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
Description
- Molded from a special high-density polymer, the cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents
- Special hinge holds base and cover together, allowing cassettes to be opened and closed repeatedly
- Cover can be removed and re-inserted easily without danger of specimen loss
- Efficient flow-through slots maximize fluid exchange and ensure proper drainage
- Covers packaged separately
Specifications
Specifications
Closure Included | No |
Color | Green |
Material | High Density Polymer |
Product Type | Tissue Cassette |