Learn More
Description
T100 BIOTUBE™ Tubes are ideal for HTLV-III testing, bacterial and hybridoma cell uptake studies.
- Polypropylene construction
- Caps sold separately
- Gross volume: 1.2 mL, with cap : 1.1 mL
- Used for procedures requiring low surface tension such as protein, nucleic acid work
- Designed to avoid potentially harmful lubricants while minimizing liquid retention
- Applications: Ideal for HTLV-III testing, bacterial and hybridoma cell uptake studies, cell harvesting, pharmaceutical quality control, receptor binding assays, RIA and EIA
Specifications
Specifications
| Autoclavable | Autoclavable |
| Capacity (Metric) | 1.2 mL |
| Material | Polypropylene |
| For Use With (Application) | Ideal for HTLV-III testing, bacterial and hybridoma cell uptake studies, cell harvesting, pharmaceutical quality control, receptor binding assays, RIA and EIA |
| For Use With (Equipment) | T100 Biotube |
| Includes | Individual Tubes without Writing Surface |
| No. per Pack | 600/Cs. |
| Sterility | Non-sterile |
| Max. RCF | 2000 x g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.