Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport™ Scientific T101 BIOTUBE™ Tube Rack

Catalog No. p-4375895 Shop All Simport Scientific Products
Click to view available options
Sterility:
Non-sterile
Sterile
Includes:
8 strips of 12 tubes
96 plain individual tubes

The T101 BIOTUBE™ Tube Rack is suitable for use with robotics systems and for transferring liquids with multichannel pipettors and auto sampling devices that conform to 96-well microplate systems.

  • Made of polypropylene
  • Conforms to SBS standard
  • Free of RNase, DNase, Pyrogen, and DNA
  • The rack has a standard microtiter footprint
  • Easy to read ID numbers and letters
  • Cover and base are keyed to prevent misalignment
  • Translucent cover
  • Volume with cap is 1.1 mL
  • Caps sold separately

Specifications

Includes 96 plain individual tubes
Product Type T101 BIOTUBE Tube Rack
Certifications/Compliance Conforms to SBS standards. RNAse, DNAse, DNA and Pyrogen free.
Autoclavable Autoclavable
Holds Microtubes (1.2 mL)
Material Polypropylene
Positions 96
Sterility Sterile
Quantity 10/Cs.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.