Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sin Nombre Virus Glycoprotein 1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2412550.025MG
Description
Sin Nombre Virus Glycoprotein 1 Polyclonal specifically detects Sin Nombre Virus Glycoprotein 1 in Virus samples. It is validated for ELISA.Specifications
Sin Nombre Virus Glycoprotein 1 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
GPC | |
Antibody was raised against a 19 amino acid peptide near the amino terminus of the Sin Nombre virus glycoprotein. The immunogen is located within amino acids 60 - 110 of Sin Nombre Virus Glycoprotein 1. Amino Acid Squence: TTTQKYNQVDWTKKSSTTE | |
Affinity Purified | |
RUO | |
Virus | |
IgG |
ELISA | |
1 mg/mL | |
ELISA 1 μg/mL | |
Envelope polyprotein, GPC, M polyprotein, SNVsMgp1 | |
Rabbit | |
126 kDa | |
0.025 mg | |
Primary | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction