Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sin Nombre Virus Glycoprotein 1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | Sin Nombre Virus Glycoprotein 1 |
---|---|
Concentration | 1 mg/mL |
Applications | ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Sin Nombre Virus Glycoprotein 1 Polyclonal specifically detects Sin Nombre Virus Glycoprotein 1 in Virus samples. It is validated for ELISA.Specifications
Sin Nombre Virus Glycoprotein 1 | |
ELISA | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
GPC | |
IgG | |
Affinity Purified |
1 mg/mL | |
Polyclonal | |
Rabbit | |
Virus | |
Envelope polyprotein, GPC, M polyprotein, SNVsMgp1 | |
Antibody was raised against a 19 amino acid peptide near the amino terminus of the Sin Nombre virus glycoprotein. The immunogen is located within amino acids 60 - 110 of Sin Nombre Virus Glycoprotein 1. Amino Acid Squence: TTTQKYNQVDWTKKSSTTE | |
Primary | |
126 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title