Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sin Nombre Virus Glycoprotein 1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB343601 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343601 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343601 Supplier Novus Biologicals Supplier No. NBP241255MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sin Nombre Virus Glycoprotein 1 Polyclonal antibody specifically detects Sin Nombre Virus Glycoprotein 1 in Virus samples. It is validated for ELISA

Specifications

Antigen Sin Nombre Virus Glycoprotein 1
Applications ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias Envelope polyprotein, GPC, M polyprotein, SNVsMgp1
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid peptide near the amino terminus of the Sin Nombre virus glycoprotein. The immunogen is located within amino acids 60 - 110 of Sin Nombre Virus Glycoprotein 1. Amino Acid Squence: TTTQKYNQVDWTKKSSTTE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 2654026
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.