Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sin1/MAPKAP1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056482T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056482T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056482T Supplier Novus Biologicals Supplier No. NB10056482SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 7 publications

Sin1/MAPKAP1 Polyclonal specifically detects Sin1/MAPKAP1 in Avian, Bovine, Canine, Chicken, Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot.

Specifications

Antigen Sin1/MAPKAP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml
Formulation PBS containing 0.2% gelatin with 0.05% Sodium Azide
Gene Accession No. Q92973
Gene Alias JC310, MEKK2-interacting protein 1, MGC2745, MIP1SIN1SIN1b, Mitogen-activated protein kinase 2-associated protein 1, mitogen-activated protein kinase associated protein 1, mSIN1, ras inhibitor MGC2745, SAPK-interacting protein 1, SIN1g, stress-activated map kinase interacting protein 1, Stress-activated map kinase-interacting protein 1, stress-activated protein kinase-interacting 1, target of rapamycin complex 2 subunit MAPKAP1, TORC2 subunit MAPKAP1
Gene Symbols MAPKAP1
Host Species Rabbit
Immunogen A portion of amino acids 85-102 (NTAQRLERLRKERQNQIK) of human Mip1 was used as the immunogen.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Autophagy, Cancer, mTOR Pathway, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 79109
Test Specificity There are multiple isoforms of this protein, ranging from approximately 310 amino acids to approximately 522 amino acids. Isoform distribution varies with tissue type.
Target Species Avian, Bovine, Rat, Canine, Chicken, Human, Mouse, Rat, Zebrafish
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.