Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIN3A Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB106993 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB106993 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB106993 Supplier Novus Biologicals Supplier No. NBP298964UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SIN3A Polyclonal specifically detects SIN3A in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SIN3A
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Formulation 50 mM Sodium Borate
Gene Alias DKFZp434K2235, FLJ90319, Histone deacetylase complex subunit Sin3a, KIAA0700, paired amphipathic helix protein Sin3a, SIN3 homolog A, transcription regulator (yeast), SIN3 homolog A, transcriptional regulator (yeast), transcriptional co-repressor Sin3A, Transcriptional corepressor Sin3a, transcriptional regulator, SIN3A
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human SIN3A.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25942
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.