Learn More
Description
Specifications
Specifications
| Antigen | SIN3A |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5 ug/ml, Flow Cytometry, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50 - 1:500, Chromatin Immunoprecipitation (ChIP) 1-3 ul |
| Formulation | PBS with 1mg/ml BSA with 0.05% Sodium Azide |
| Gene Accession No. | Q96ST3, Q60520 |
| Gene Alias | DKFZp434K2235, FLJ90319, Histone deacetylase complex subunit Sin3a, KIAA0700, paired amphipathic helix protein Sin3a, SIN3 homolog A, transcription regulator (yeast), SIN3 homolog A, transcriptional regulator (yeast), transcriptional co-repressor Sin3A, Transcriptional corepressor Sin3a, transcriptional regulator, SIN3A |
| Gene Symbols | SIN3A |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
