Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Single Glass Cone
SCI_ED

Catalog No. S89366 Shop All Eisco Products
Click to view available options
Standard Taper Joint No.:
10/19
12/21
14/23
19/26
24/29
29/32
34/35

A variety of standard cone sizes are available, each with a minimum shank length of 100 mm

  • Single cones used to connect glassware
  • Made from high quality borosilicate 3.3 glass
  • Cone features ground glass
  • Autoclavable
  • Warranty and Services

    1 year limited manufacturer warranty

    Specifications

    Cone Size 10/19
    For Use With (Application) Connect glassware
    Height (Metric) 10.6 cm
    Length (Metric) 2 cm
    Material Borosilicate Glass (3.3)
    Product Type Single Cone
    Quantity 1
    Standard Taper Joint No. 10/19
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.