Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIPA1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB342145 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342145 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342145 Supplier Novus Biologicals Supplier No. NBP281850MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SIPA1 Polyclonal antibody specifically detects SIPA1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen SIPA1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias GTPase-activating protein Spa-1, MGC102688, MGC17037, p130 SPA-1, signal-induced proliferation-associated 1, signal-induced proliferation-associated protein 1, sipa-1, SPA1signal-induced proliferation-associated gene 1
Host Species Rabbit
Immunogen SIPA1 antibody was raised against a 19 amino acid synthetic peptide from near the amino terminus of human SIPA1. The immunogen is located within amino acids 40 - 90 of SIPA1. Amino Acid Squence: DAGEARPPTPASPRARAHS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6494
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.