Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIPA1L2 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB356639 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB356639 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB356639 Supplier Novus Biologicals Supplier No. NBP177090PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SIPA1L2 Polyclonal antibody specifically detects SIPA1L2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen SIPA1L2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias FLJ23632, signal-induced proliferation-associated 1 like 2, signal-induced proliferation-associated 1-like protein 2, SIPA1-like protein 2, SPA-1-like 2, SPAL2
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid synthetic peptide from near the amino terminus of human SIPA1L2. The immunogen is located within the first 50 amino acids of SIPA1L2. Amino Acid Squence: SKFKDPPRIMQSDDYFARK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 57568
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.