Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIPA1L3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP230363 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP230363 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP230363 Supplier Novus Biologicals Supplier No. NBP230363
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SIPA1L3 Polyclonal specifically detects SIPA1L3 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).

Specifications

Antigen SIPA1L3
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2 - 10 ug/mg lysate, Western Blot (Negative)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_055888.1
Gene Alias KIAA0545, Signal-Induced Proliferation-Associated 1 Like 3, Signal-Induced Proliferation-Associated 1-Like Protein 3, SIPA1-Like Protein 3, SPA-1-Like 3, SPA-1-Like Protein 3, SPAL3
Gene Symbols SIPA1L3
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1500 to 1550 of human Signal-Induced Proliferation-Associated 1 Like 3 using the numbering given in entry NP_055888.1 (GeneID 23094).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23094
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.