Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIPA1L3 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB290162 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290162 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290162 Supplier Novus Biologicals Supplier No. NBP230362PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SIPA1L3 Polyclonal antibody specifically detects SIPA1L3 in Human samples. It is validated for Western Blot, Immunoprecipitation (Negative)

Specifications

Antigen SIPA1L3
Applications Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunoprecipitation (Negative)
Formulation PBS
Gene Alias KIAA0545, Signal-Induced Proliferation-Associated 1 Like 3, Signal-Induced Proliferation-Associated 1-Like Protein 3, SIPA1-Like Protein 3, SPA-1-Like 3, SPA-1-Like Protein 3, SPAL3
Host Species Rabbit
Immunogen Maps to a region between residue 1 to 50 of human Signal-Induced Proliferation-Associated 1 Like 3 using the numbering given in entry NP_055888.1 (GeneID 23094).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 23094
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.