Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Sipatrigine
GREENER_CHOICE

Catalog No. 404410
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

NaV blocker; neuroprotective

Blocker of voltage-dependent sodium channels (NaV). Inhibits glutamate release; displays neuroprotective activity in rat models of cerebral ischemia. Also thought to block Ca2+ channels. Analog of lamotrigine (Cat. No. 1611).

Chemical Identifiers

CAS 130800-90-7
Molecular Formula C15H16Cl3N5

Specifications

Quantity 10 mg
Formula Weight 372.68
Purity >99%
Molecular Weight (g/mol) 372.678
Chemical Name or Material Sipatrigine
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.