Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sir2.1 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001923 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001923 0.1 mL
NB1001923SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1001923 Supplier Novus Biologicals Supplier No. NB1001923
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

Sir2.1 Polyclonal specifically detects Sir2.1 in C. elegans samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen Sir2.1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:3000, Immunohistochemistry 1:200-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:500
Formulation Whole antisera with 0.05% Sodium Azide
Gene Alias EC 3.5.1, EC 3.5.1.-, FLJ35621, FLJ37491, S.cerevisiae, homolog) 2, sirtuin 2, sirtuin type 2
Gene Symbols SIRT2
Host Species Rabbit
Immunogen A full-length recombinant protein to Sir2.1.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Cytoskeleton Markers, DNA Repair, Neuronal Cell Markers, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 22933
Test Specificity C. elegans.
Target Species C. elegans
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.