Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sir2.1 Rabbit anti-Human, Clone: SN70-04, Novus Biologicals™
SDP

Catalog No. NBP267707 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267707 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267707 Supplier Novus Biologicals Supplier No. NBP267707

Rabbit Monoclonal Antibody

Sir2.1 Monoclonal specifically detects Sir2.1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen Sir2.1
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone SN70-04
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias EC 3.5.1, EC 3.5.1.-, FLJ35621, FLJ37491, S.cerevisiae, homolog) 2, sirtuin 2, sirtuin type 2
Gene Symbols SIRT2
Host Species Rabbit
Immunogen Synthetic peptide within Human Sir2.1 aa 335-389/ 389. (SwissProt: Q8IXJ6 Human; SwissProt: Q5RJQ4 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Cancer, Chromatin Research, Cytoskeleton Markers, DNA Repair, Neuronal Cell Markers, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 22933
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.