Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRP alpha/CD172a Antibody (602411), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB369840 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB369840 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB369840 Supplier Novus Biologicals Supplier No. FAB4546J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SIRP alpha/CD172a Monoclonal specifically detects SIRP alpha/CD172a in Human samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry, Western Blot.

Specifications

Antigen SIRP alpha/CD172a
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 602411
Conjugate Janelia Fluor 646
Dilution Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Bit, BITbrain-immunoglobulin-like molecule with tyrosine-based activation motifs, Brain Ig-like molecule with tyrosine-based activation motifs, CD172 antigen-like family member A, CD172a, CD172a antigen, Inhibitory receptor SHPS-1, Macrophage fusion receptor, MFRtyrosine phosphatase SHP substrate 1, MYD1, MYD-1, MyD-1 antigen, P84, protein tyrosine phosphatase, non-receptor type substrate 1, PTPNS1, SHP substrate 1, SHPS-1, SHPS1CD172A, signal-regulatory protein alpha, Signal-regulatory protein alpha-1, Signal-regulatory protein alpha-2, Signal-regulatory protein alpha-3, SIRPalpha, SIRP-ALPHA-1, Sirp-alpha-2, SIRPalpha2, Sirp-alpha-3, SIRPtyrosine-protein phosphatase non-receptor type substrate 1
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human SIRP alpha/CD172a, Gly27-Asn370 (predicted), Accession # P78324
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Protein Phosphatase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 140885
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.