Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRP alpha/CD172a Antibody (OTI7B3), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N274197J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274197J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274197J646 Supplier Novus Biologicals Supplier No. NBP274197JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SIRP alpha/CD172a Monoclonal specifically detects SIRP alpha/CD172a in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen SIRP alpha/CD172a
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI7B3
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Bit, BITbrain-immunoglobulin-like molecule with tyrosine-based activation motifs, Brain Ig-like molecule with tyrosine-based activation motifs, CD172 antigen-like family member A, CD172a, CD172a antigen, Inhibitory receptor SHPS-1, Macrophage fusion receptor, MFRtyrosine phosphatase SHP substrate 1, MYD1, MYD-1, MyD-1 antigen, P84, protein tyrosine phosphatase, non-receptor type substrate 1, PTPNS1, SHP substrate 1, SHPS-1, SHPS1CD172A, signal-regulatory protein alpha, Signal-regulatory protein alpha-1, Signal-regulatory protein alpha-2, Signal-regulatory protein alpha-3, SIRPalpha, SIRP-ALPHA-1, Sirp-alpha-2, SIRPalpha2, Sirp-alpha-3, SIRPtyrosine-protein phosphatase non-receptor type substrate 1
Gene Symbols SIRPA
Host Species Mouse
Immunogen Full length recombinant protein of human SIRP alpha/CD172a (NP_001035111) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline GPCR, Protein Phosphatase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 140885.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.