Learn More
SIRP alpha/CD172a Rabbit anti-Human, Clone: S08-1G4, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | SIRP alpha/CD172a |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Clone | S08-1G4 |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Immunocytochemistry/Immunofluorescence |
| Formulation | 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA |
| Gene Alias | Bit, BITbrain-immunoglobulin-like molecule with tyrosine-based activation motifs, Brain Ig-like molecule with tyrosine-based activation motifs, CD172 antigen-like family member A, CD172a, CD172a antigen, Inhibitory receptor SHPS-1, Macrophage fusion receptor, MFRtyrosine phosphatase SHP substrate 1, MYD1, MYD-1, MyD-1 antigen, P84, protein tyrosine phosphatase, non-receptor type substrate 1, PTPNS1, SHP substrate 1, SHPS-1, SHPS1CD172A, signal-regulatory protein alpha, Signal-regulatory protein alpha-1, Signal-regulatory protein alpha-2, Signal-regulatory protein alpha-3, SIRPalpha, SIRP-ALPHA-1, Sirp-alpha-2, SIRPalpha2, Sirp-alpha-3, SIRPtyrosine-protein phosphatase non-receptor type substrate 1 |
| Host Species | Rabbit |
| Immunogen | Recombinant protein of human SIRP alpha/CD172a (Uniprot#: P78324) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.