Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIRP alpha/CD172a Rabbit anti-Human, Clone: S08-1G4, Novus Biologicals™

Rabbit Monoclonal Antibody
$263.00 - $573.00
Specifications
Antigen | SIRP alpha/CD172a |
---|---|
Clone | S08-1G4 |
Dilution | Western Blot, Immunocytochemistry/Immunofluorescence |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Description
SIRP alpha/CD172a Monoclonal antibody specifically detects SIRP alpha/CD172a in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
SIRP alpha/CD172a | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Bit, BITbrain-immunoglobulin-like molecule with tyrosine-based activation motifs, Brain Ig-like molecule with tyrosine-based activation motifs, CD172 antigen-like family member A, CD172a, CD172a antigen, Inhibitory receptor SHPS-1, Macrophage fusion receptor, MFRtyrosine phosphatase SHP substrate 1, MYD1, MYD-1, MyD-1 antigen, P84, protein tyrosine phosphatase, non-receptor type substrate 1, PTPNS1, SHP substrate 1, SHPS-1, SHPS1CD172A, signal-regulatory protein alpha, Signal-regulatory protein alpha-1, Signal-regulatory protein alpha-2, Signal-regulatory protein alpha-3, SIRPalpha, SIRP-ALPHA-1, Sirp-alpha-2, SIRPalpha2, Sirp-alpha-3, SIRPtyrosine-protein phosphatase non-receptor type substrate 1 | |
Recombinant protein of human SIRP alpha/CD172a (Uniprot#: P78324) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
S08-1G4 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
GPCR, Protein Phosphatase, Signal Transduction | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
140885 | |
IgG | |
Affinity purified | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title