Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRP beta 1/gamma Antibody (308906), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB313776 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB313776 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB313776 Supplier Novus Biologicals Supplier No. FAB2096APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SIRP beta 1/gamma Monoclonal antibody specifically detects SIRP beta 1/gamma in Human samples. It is validated for Flow Cytometry

Specifications

Antigen SIRP beta 1/gamma
Applications Flow Cytometry
Classification Monoclonal
Clone 308906
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD172b;SIRPB1;SIRP-BETA-1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human SIRP beta 1, Gly26-Ala369 (Arg53His and Ala363Pro), Accession # O00241.4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Signaling, Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.