Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRP beta 1/gamma Antibody (OX117), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB015369 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB015369 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB015369 Supplier Novus Biologicals Supplier No. NBP281071AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SIRP beta 1/gamma Monoclonal specifically detects SIRP beta 1/gamma in Human samples. It is validated for Flow Cytometry, ELISA.

Specifications

Antigen SIRP beta 1/gamma
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone OX117
Conjugate Alexa Fluor 750
Dilution Flow Cytometry, ELISA
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Recombinant human SIRPgamma:CD4 fusion protein.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody binds to human SIRP beta 1/gamma.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.