Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRP delta Antibody (2588A), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. NB315366 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB315366 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB315366 Supplier Novus Biologicals Supplier No. FAB10138PECY55
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SIRP delta Monoclonal antibody specifically detects SIRP delta in Human samples. It is validated for Flow Cytometry

Specifications

Antigen SIRP delta
Applications Flow Cytometry
Classification Monoclonal
Clone 2588A
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS
Gene Alias dJ576H24.4, Protein tyrosine phosphatase non-receptor type substrate 1-like 2, protein tyrosine phosphatase, non-receptor type substrate 1-like 2, PTPNS1L2, signal-regulatory protein delta, SIRP-delta
Host Species Rabbit
Immunogen Human embryonic kidney cell, HEK293-derived human SIRP delta, Phe30-Arg197, Accession # Q9H106
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 128646
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.