Learn More
SIRT1 Monoclonal Antibody (2G1/F7), Active Motif™

Description
Specifications
Specifications
| Antigen | SIRT1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 2G1/F7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 10mM sodium phosphate with 150mM NaCl, 30% glycerol and 0.035% sodium azide; pH 7.5 |
| Gene | SIRT1 |
| Gene Accession No. | Q96EB6 |
| Gene Alias | 75SirT1; AA673258; hSIR2; hSIRT1; mSIR2a; NAD-dependent deacetylase sirtuin-1; NAD-dependent protein deacetylase sirtuin-1; OTTHUMP00000198111; OTTHUMP00000198112; Regulatory protein SIR2 homolog 1; RP11-57G10.3; Sir2; Sir2a; SIR2alpha; Sir2l1; sir2-like 1; SIR2-like protein 1; SIRT1; SirtT1 75 kDa fragment; sirtuin 1; sirtuin 1 ((silent mating type information regulation 2, homolog) 1; sirtuin type 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.