Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRT4 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB056045 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB056045 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB056045 Supplier Novus Biologicals Supplier No. NBP298856UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SIRT4 Polyclonal specifically detects SIRT4 in Human samples. It is validated for Western Blot.

Specifications

Antigen SIRT4
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EC 2.4.2.-, EC 3.5.1, MGC130046, MGC130047, MGC57437, SIR2L4NAD-dependent ADP-ribosyltransferase sirtuin-4, sir2-like 4, SIR2-like protein 4, sirtuin (silent mating type information regulation 2 homolog) 4 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S. cerevisiae, homolog) 4, sirtuin 4, sirtuin type 4
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human SIRT4.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, Epigenetics, Homologous Recombination
Primary or Secondary Primary
Gene ID (Entrez) 23409
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.