Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRT6 Antibody (OTI1G3), FITC, Novus Biologicals™
SDP

Catalog No. NBP274198F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274198F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274198F Supplier Novus Biologicals Supplier No. NBP274198F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SIRT6 Monoclonal specifically detects SIRT6 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SIRT6
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1G3
Conjugate FITC
Dilution Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 3.5.1.-, NAD-dependent deacetylase sirtuin-6, SIR2L6sir2-related protein type 6, SIR2-like protein 6, sirtuin (silent mating type information regulation 2 homolog) 6 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S. cerevisiae, homolog) 6, sirtuin 6, sirtuin type 6
Gene Symbols SIRT6
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 88-352 of human SIRT6 (NP_057623) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Chromatin Modifiers, Chromatin Research, DNA Repair, Epigenetics, Histone Deacetylases, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51548.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.