Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRT6 Rabbit anti-Human, Janelia Fluor 549, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB056069 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB056069 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB056069 Supplier Novus Biologicals Supplier No. NBP298858JF549

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

SIRT6 Polyclonal specifically detects SIRT6 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SIRT6
Applications Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias EC 3.5.1.-, NAD-dependent deacetylase sirtuin-6, SIR2L6sir2-related protein type 6, SIR2-like protein 6, sirtuin (silent mating type information regulation 2 homolog) 6 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S. cerevisiae, homolog) 6, sirtuin 6, sirtuin type 6
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human SIRT6.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, DNA Repair, Epigenetics, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51548
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.