Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRT7 Antibody (1E2B2) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP252564 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP252564 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP252564 Supplier Novus Biologicals Supplier No. NBP252564
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SIRT7 Monoclonal specifically detects SIRT7 in Human samples. It is validated for CyTOF, ELISA, Flow Cytometry, Western Blot.

Specifications

Antigen SIRT7
Applications CyTOF, ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 1E2B2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 3.5.1.-, MGC126840, MGC126842, silent mating type information regulation 2, S.cerevisiae, homolog 7, SIR2L7NAD-dependent deacetylase sirtuin-7, SIR2-like protein 7, sir2-related protein type 7, sirtuin (silent mating type information regulation 2 homolog) 7 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S.cerevisiae, homolog) 7, sirtuin 7, sirtuin type 7
Gene Symbols SIRT7
Host Species Mouse
Immunogen Purified recombinant fragment of human SIRT7 (AA: 1-105) expressed in E. Coli.
Molecular Weight of Antigen 44.9 kDa
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline DNA Repair, Epigenetics, Histone Deacetylases
Primary or Secondary Primary
Gene ID (Entrez) 51547
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.