Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIRT7 Antibody

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11081744
Description
SIRT7 Polyclonal specifically detects SIRT7 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
SIRT7 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide | |
EC 3.5.1.-, MGC126840, MGC126842, silent mating type information regulation 2, S.cerevisiae, homolog 7, SIR2L7NAD-dependent deacetylase sirtuin-7, SIR2-like protein 7, sir2-related protein type 7, sirtuin (silent mating type information regulation 2 homolog) 7 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S.cerevisiae, homolog) 7, sirtuin 7, sirtuin type 7 | |
Rabbit | |
45 kDa | |
0.1 mL | |
DNA Repair, Epigenetics, Histone Deacetylases | |
51547 | |
Human, Mouse | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:20 - 1:100 | |
Q9NRC8 | |
SIRT7 | |
Synthetic peptide made to an internal portion of human SIRT7 (within residues 100-200). [Swiss-Prot# Q9NRC8] | |
Affinity Purified | |
RUO | |
Primary | |
Human and mouse. Immunogen sequence has 88% homology to bovine and porcine and 82% homology to Drosophila. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction