Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIRT7 Antibody

Rabbit Polyclonal Antibody
$485.00
Specifications
Antigen | SIRT7 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:20 - 1:100 |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11081744
![]() |
Novus Biologicals
NB11081744 |
0.1 mL |
Each of 1 for $485.00
|
|
|||||
Description
SIRT7 Polyclonal specifically detects SIRT7 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
SIRT7 | |
Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:20 - 1:100 | |
Polyclonal | |
Rabbit | |
DNA Repair, Epigenetics, Histone Deacetylases | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide | |
EC 3.5.1.-, MGC126840, MGC126842, silent mating type information regulation 2, S.cerevisiae, homolog 7, SIR2L7NAD-dependent deacetylase sirtuin-7, SIR2-like protein 7, sir2-related protein type 7, sirtuin (silent mating type information regulation 2 homolog) 7 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S.cerevisiae, homolog) 7, sirtuin 7, sirtuin type 7 | |
SIRT7 | |
IgG | |
Affinity Purified | |
Human and mouse. Immunogen sequence has 88% homology to bovine and porcine and 82% homology to Drosophila. |
1 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human, Mouse | |
Q9NRC8 | |
51547 | |
Synthetic peptide made to an internal portion of human SIRT7 (within residues 100-200). [Swiss-Prot# Q9NRC8] | |
Primary | |
Store at 4C. Do not freeze. | |
45 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title