Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sirtuin 1/SIRT1 Antibody (1F3) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB014022 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB014022 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB014022 Supplier Novus Biologicals Supplier No. NBP280949
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Sirtuin 1/SIRT1 Monoclonal specifically detects Sirtuin 1/SIRT1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), Immunohistochemistry Free-Floating, CyTOF-ready.

Specifications

Antigen Sirtuin 1/SIRT1
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Free Floating), CyTOF
Classification Monoclonal
Clone 1F3
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1:500-1:2000, Flow Cytometry : 1 ug per million cells, ELISA : 1:10000, Immunohistochemistry : 1:200-1:1000, Immunocytochemistry/Immunofluorescence : 1:10-1:1000, Immunohistochemistry-Paraffin : 1:200-1:1000, Flow (Intracellular), Immunohistochemistry Free-Floating reported in scientific literature (PMID 26327687), CyTOF-ready
Formulation PBS
Gene Alias EC 3.5.1, S. cerevisiae, homolog) 1, sirtuin 1
Gene Symbols SIRT1
Host Species Mouse
Immunogen Purified recombinant fragment of human SIRT1 (amino acids: 265-452) expressed in E. coli. [UniProt# Q96EB6]
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Chromatin Research, Cognition and Behavior, DNA Repair, Epigenetics, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 23411
Test Specificity Human, mouse, rat and monkey.
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.