Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sirtuin 1/SIRT1 Antibody (1F3) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP151641 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP151641 0.1 mL
NBP151641SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP151641 Supplier Novus Biologicals Supplier No. NBP151641
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 5 publications

Sirtuin 1/SIRT1 Monoclonal specifically detects Sirtuin 1/SIRT1 in Human, Mouse, Rat, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), Immunohistochemistry Free-Floating.

Specifications

Antigen Sirtuin 1/SIRT1
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1F3
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1 ug per million cells, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, Flow (Intracellular), Immunohistochemistry Free-Floating reported in scientific literature (PMID 26327687)
Gene Accession No. Q96EB6
Gene Alias EC 3.5.1, S. cerevisiae, homolog) 1, sirtuin 1
Gene Symbols SIRT1
Host Species Mouse
Immunogen Purified recombinant fragment of human SIRT1 (amino acids: 265-452) expressed in E. coli. [UniProt# Q96EB6]
Molecular Weight of Antigen 120 kDa
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Chromatin Research, DNA Repair, Epigenetics, Histone Deacetylases
Primary or Secondary Primary
Gene ID (Entrez) 23411
Target Species Human, Mouse, Rat, Primate, Rabbit
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Precipitation
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.