Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sirtuin 1/SIRT1 Antibody (1F3) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 5 publications
$192.47 - $493.10
Specifications
| Antigen | Sirtuin 1/SIRT1 |
|---|---|
| Clone | 1F3 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1 ug per million cells, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, Flow (Intracellular), Immunohistochemistry Free-Floating reported in scientific literature (PMID 26327687) |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP151641SS
![]() |
Novus Biologicals
NBP151641SS |
0.025 mL |
Each for $192.47
|
|
|||||
NBP151641
![]() |
Novus Biologicals
NBP151641 |
0.1 mL |
Each for $493.10
|
|
|||||
Description
Sirtuin 1/SIRT1 Monoclonal specifically detects Sirtuin 1/SIRT1 in Human, Mouse, Rat, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), Immunohistochemistry Free-Floating.Specifications
| Sirtuin 1/SIRT1 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Chromatin Research, DNA Repair, Epigenetics, Histone Deacetylases | |
| Q96EB6 | |
| 23411 | |
| Purified recombinant fragment of human SIRT1 (amino acids: 265-452) expressed in E. coli. [UniProt# Q96EB6] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1F3 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1 ug per million cells, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, Flow (Intracellular), Immunohistochemistry Free-Floating reported in scientific literature (PMID 26327687) | |
| Monoclonal | |
| Precipitation | |
| RUO | |
| Human, Mouse, Rat, Primate, Rabbit | |
| EC 3.5.1, S. cerevisiae, homolog) 1, sirtuin 1 | |
| SIRT1 | |
| IgG1 | |
| Ammonium sulfate precipitation | |
| 120 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title