Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sirtuin 1/SIRT1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227205 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227205 0.1 mg
NBP227205SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227205 Supplier Novus Biologicals Supplier No. NBP227205
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 7 publications

Sirtuin 1/SIRT1 Polyclonal specifically detects Sirtuin 1/SIRT1 in Human, Mouse, Porcine, Amphibian, Bovine, Canine, Chicken, Chinese Hamster, Equine, Feline, Hamster, Primate, Rabbit samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen Sirtuin 1/SIRT1
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-4ug/ml, Simple Western 1:100
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 3.5.1, S. cerevisiae, homolog) 1, sirtuin 1
Gene Symbols SIRT1
Host Species Rabbit
Immunogen A portion of amino acids 400-450 from human SIRT1 was used as the immunogen for this antibody.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Chromatin Research, DNA Repair, Epigenetics, Histone Deacetylases, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 23411
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, mouse, rat, orangutan, chimpanzee, panda, Carolina anole, cow, new world monkey, dog, Chinese hamster, horse, chicken, mole rat, rhesus monkey, wild turkey, opossum, platypus, rabbit, pig, zebra finch, frog, cat, and white-cheeked gibbon, and 89% homologous in zebrafish, cichlid fish, and killifish.
Target Species Human, Mouse, Pig, Amphibian, Bovine, Canine, Chicken, Chinese Hamster, Equine, Feline, Hamster, Primate, Rabbit
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.