Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sirtuin 5/SIRT5 Antibody (OTI2F1), FITC, Novus Biologicals™
SDP

Catalog No. NBP274199F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274199F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274199F Supplier Novus Biologicals Supplier No. NBP274199F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Sirtuin 5/SIRT5 Monoclonal specifically detects Sirtuin 5/SIRT5 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated.

Specifications

Antigen Sirtuin 5/SIRT5
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay
Classification Monoclonal
Clone OTI2F1
Conjugate FITC
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 3.5.1, EC 3.5.1.-, FLJ36950, NAD-dependent deacetylase sirtuin-5, silent mating type information regulation 2, S.cerevisiae, homolog 5, SIR2L5, sir2-like 5, SIR2-like protein 5, sirtuin (silent mating type information regulation 2 homolog) 5 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S.cerevisiae, homolog) 5, sirtuin 5, sirtuin type 5
Gene Symbols SIRT5
Host Species Mouse
Immunogen Full length recombinant protein of human Sirtuin 5/SIRT5 (NP_036373) produced in HEK293 cell.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Epigenetics, Histone Deacetylases
Primary or Secondary Primary
Gene ID (Entrez) 23408.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.