Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIT1 Antibody (SIT-01), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB500486X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500486X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500486X Supplier Novus Biologicals Supplier No. NB500486AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SIT1 Monoclonal specifically detects SIT1 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen SIT1
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone SIT-01
Conjugate Alexa Fluor 488
Dilution Western Blot, Flow Cytometry
Gene Alias gp30/40, MGC125908, MGC125909, RP11-331F9.5, SHP2 interacting transmembrane adaptor, SHP2-interacting transmembrane adapter protein, SHP2-interacting transmembrane adaptor protein, signaling threshold regulating transmembrane adaptor 1, signaling threshold-regulating transmembrane adapter 1, SITMGC125910, suppression inducing transmembrane adaptor 1, Suppression-inducing transmembrane adapter 1
Gene Symbols SIT1
Host Species Mouse
Immunogen Bacterially produced recombinant intracellular fragment of human SIT.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 27240
Test Specificity The antibody SIT-01 reacts with SHP2-interacting transmembrane adaptor protein (SIT) expressed exclusively in lymphoid organs.
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.